missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexafluorosilicate, 99.999%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder and/or Chunks |
| Quantity | 10 g |
| Assay Percent Range | 99.999% |
| Linear Formula | (NH4)2SiF6 |
| Merck Index | 15,524 |
| Solubility Information | Solubility in water: 18.6 g/100mL (17°C). |
| Formula Weight | 178.15 |
| Percent Purity | 99.999% |
| Grade | Trace Metal Basis |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protectio |
| EINECSNumber | 240-968-3 |
RUO – Research Use Only