missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexafluorophosphate, 99%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 198°C |
| Color | White to Beige |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 5 g |
| Assay Percent Range | 99% |
| Linear Formula | NH4PF6 |
| Merck Index | 15, 523 |
| Solubility Information | Solubility in water: 74.8g/100mL (20 c). Other solubilities: soluble in acetone,methanol,ethanol and methyl,acetate |
| Formula Weight | 163 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement: Causes severe skin burns and eye damage. Causes serious eye damage. |
| missing translation for 'healthHazard3' | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 241-009-1 |
| TSCA | TSCA |
RUO – Research Use Only