missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorogermanate, 99.99% min (metals basis)

CAS: 16962-47-3 | H8F6GeN2

Supplier:  Thermo Scientific Chemicals 04317906

 View more versions of this product

Catalog No. AA4317906


Only null left
Explore available promotions

Description

Description

Ammonium hexafluorogermanate is used to substitutes Al with Ge in Y type zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium hexafluorogermanate
16962-47-3
5 g
H8F6GeN2
MFCD00010880
Soluble in water.
≥99.99%
White
Crystalline
(metals basis)
(NH4)2GeF6
UN3288
222.71
2.564 g/mL

Chemical Identifiers

16962-47-3
H8F6GeN2
MFCD00010880
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 241-037-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only