missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluoroaluminate, 98%

CAS: 7784-19-2 | AlF6H12N3 | 195.09 g/mol

$76.26 - $236.55

Chemical Identifiers

CAS 7784-19-2
Molecular Formula AlF6H12N3
Molecular Weight (g/mol) 195.09
MDL Number MFCD00042144
InChI Key OYHBNKHFKHBTRQ-UHFFFAOYSA-K
Synonym ammonium hexafluoroaluminate, ammonium cryolite, ammonium fluoaluminate, ammonium aluminum fluoride, triammonium hexafluoroaluminate, triammonium aluminum hexafluoride, triammoniumhexafluoroaluminate 3-, hexafluoroaluminate 3-triammonium, aluminum ammonium fluoride al nh4 3f6, ammonium hexafluoroaluminate nh4 3alf6
PubChem CID 24565
IUPAC Name triazanium;hexafluoroaluminum(3-)
SMILES [NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2058209
View Documents
Thermo Scientific Chemicals
B2058209
10 g
Each for $76.26
Only null left
 
AAB2058218
View Documents
Thermo Scientific Chemicals
B2058218
50 g
Each for $236.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7784-19-2
195.09
OYHBNKHFKHBTRQ-UHFFFAOYSA-K
24565
[NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F
AlF6H12N3
MFCD00042144
ammonium hexafluoroaluminate, ammonium cryolite, ammonium fluoaluminate, ammonium aluminum fluoride, triammonium hexafluoroaluminate, triammonium aluminum hexafluoride, triammoniumhexafluoroaluminate 3-, hexafluoroaluminate 3-triammonium, aluminum ammonium fluoride al nh4 3f6, ammonium hexafluoroaluminate nh4 3alf6
triazanium;hexafluoroaluminum(3-)

Specifications

7784-19-2
AlF6H12N3
UN3288
ammonium hexafluoroaluminate, ammonium cryolite, ammonium fluoaluminate, ammonium aluminum fluoride, triammonium hexafluoroaluminate, triammonium aluminum hexafluoride, triammoniumhexafluoroaluminate 3-, hexafluoroaluminate 3-triammonium, aluminum ammonium fluoride al nh4 3f6, ammonium hexafluoroaluminate nh4 3alf6
[NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F
195.09
195.09
Odorless
10 g
MFCD00042144
14,524
OYHBNKHFKHBTRQ-UHFFFAOYSA-K
triazanium;hexafluoroaluminum(3-)
24565
98%
Ammonium hexafluoroaluminate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 232-052-7

RTECSNumber : BD0050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only