missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), 99.9% (metals basis), Os 42.5% min

Catalog No. AA4287403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4287403 1 g
AA4287406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4287403 Supplier Thermo Scientific Chemicals Supplier No. 04287403

CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol

Ammonium hexachloroosmate(IV) acts as a pharmaceutical, organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12125-08-5
Molecular Formula Cl6H8N2Os
Molecular Weight (g/mol) 439.00
MDL Number MFCD00010883
InChI Key JRHMYOOMZKAPKT-UHFFFAOYSA-J
Synonym ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
PubChem CID 11729867
IUPAC Name diazanium;hexachloroosmium(2-)
SMILES [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 170° (sublimation)
Color Brown to Black
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.9% (Os 42.5% min.) (Metals basis)
Solubility Information Soluble in water.
Formula Weight 438.99
Sensitivity Hygroscopic
Density 2.9
Chemical Name or Material Ammonium hexachloroosmate(IV)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-188-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only