missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Formate, LC-MS Ultra, Solstice
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Linear Formula | HCO2NH4 |
| UN Number | NONH for all modes of transport |
| Beilstein | 3625095 |
| Formula Weight | 63.06g/mol |
| Grade | LC-MS Ultra |
| Density | 1.26g/mL at 25°C |
| Chemical Name or Material | Ammonium Formate |
Safety and Handling
| EINECSNumber | 208-753-9 |
| RTECSNumber | BQ6650000 |