missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, Crystal, 97%, Spectrum™ Chemical

Catalog No. p-7164992
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608657 500 g Poly Bottle
18608653 1 kg Poly Bottle
18608655 2.5 kg Poly Pail
18608654 12 kg Poly Pail
18608656 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608657 Supplier Spectrum Chemical Mfg Cor Supplier No. A1190500GM
Only null left

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.06
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
IUPAC Name formic acid amine
SMILES N.OC=O

Specifications

pH 5.5 to 7.5
CAS Max % 1
Quantity 500 g
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Residue after Ignition 0.0001

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.