missing translation for 'onlineSavingsMsg'
Learn More

Ammonium formate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 168610051

Catalog No. AC168610051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ammonium formate
5kg
CH5NO2
02,8
15,52
Solubility in water: soluble. Other solubilities: soluble in alcohol
C(=O)[O-].[NH4+]
63.06
CHEBI:63050
97%
Plastic drum
540-69-2
97%
HCO2NH4
01,38; 13,230; 15,13
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
VZTDIZULWFCMLS-UHFFFAOYSA-N
azanium;formate
2723923
63.06
Authentic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protect

GHS Signal Word: Warning

EINECSNumber : 208-753-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only