missing translation for 'onlineSavingsMsg'
Learn More

Ammonium formate, 97%

CAS: 540-69-2 | CH5NO2 | 63.06 g/mol

Supplier:  Thermo Scientific Chemicals 168610010

 View more versions of this product

Catalog No. AC168610010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium formate
540-69-2
6 to 7 (5% soln. at 20°C)
1 kg
CH5NO2
02,8
15,52
Solubility in water: soluble. Other solubilities: soluble in alcohol
C(=O)[O-].[NH4+]
63.06
CHEBI:63050
97%
Plastic bottle
115.0°C to 120.0°C
White
Crystalline Powder and/or Crystals
97%
HCO2NH4
01,38; 13,230; 15,13
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
VZTDIZULWFCMLS-UHFFFAOYSA-N
azanium;formate
2723923
63.06
Authentic

Chemical Identifiers

540-69-2
63.06
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
CHEBI:63050
C(=O)[O-].[NH4+]
CH5NO2
VZTDIZULWFCMLS-UHFFFAOYSA-N
2723923
azanium;formate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protect

GHS Signal Word: Warning

EINECSNumber : 208-753-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only