missing translation for 'onlineSavingsMsg'
Learn More

Ammonium formate, 97%

CAS: 540-69-2 | CH5NO2 | 63.056 g/mol

Supplier:  Thermo Scientific Chemicals A106990B

 View more versions of this product

Catalog No. AAA106990B


Only null left
Explore available promotions

Description

Description

Ammonium formate is widely used in various organic reactions like Leuckart reaction which involves the reductive amination of aldehydes and ketones. It serves as a buffer in high performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC/MS). It finds application in palladium on carbon (Pd/C) reduction of functional groups. For example, reduction of alkenes to alkanes and formaldehyde to methanol. It is also used to prepare formic acid insitu as well as used to store formic acid by making it as an ammonium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium formate is widely used in various organic reactions like Leuckart reaction which involves the reductive amination of aldehydes and ketones. It serves as a buffer in high performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC/MS). It finds application in palladium on carbon (Pd/C) reduction of functional groups. For example, reduction of alkenes to alkanes and formaldehyde to methanol. It is also used to prepare formic acid insitu as well as used to store formic acid by making it as an ammonium salt.

Solubility
Soluble in water, liquid ammonia and alcohol.

Notes
Hygroscopic. Incompatible with strong acids and strong oxidizing agents.
Specifications

Specifications

Ammonium formate
540-69-2
1.28
CH5NO2
MFCD00013103
3625095
14,523
Soluble in water,liquid ammonia and alcohol.
C(=O)[O-].[NH4+]
63.056
CHEBI:63050
97%
Water <3%
∼116°C
Ammonia-like
HCO2NH4
1000 g
Hygroscopic
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
VZTDIZULWFCMLS-UHFFFAOYSA-N
azanium;formate
2723923
63.06

Chemical Identifiers

540-69-2
63.056
VZTDIZULWFCMLS-UHFFFAOYSA-N
2723923
azanium;formate
CH5NO2
MFCD00013103
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
CHEBI:63050
C(=O)[O-].[NH4+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 208-753-9

RTECSNumber : BQ6650000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only