missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoborate, Crystal, Technical, Spectrum™ Chemical

Catalog No. 18608637
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 lb.
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608637 125 g Poly Bottle
18608638 500 g Poly Bottle
18608640 50 lb. Poly Bottle
18608639 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608637 Supplier Spectrum Chemical Mfg Cor Supplier No. A1180125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 13826-83-0

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 125 g
Molecular Weight (g/mol) 104.84
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Ammonium Fluoborate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.