Learn More
Ammonium dihydrogen phosphate, ACS, 98.0% min
Description
Preparation of baking powder with sodium bicarbonate, in fermentation.Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 190°C |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Assay Percent Range | 98% min. |
| Solubility Information | Freely soluble in water. Slightly soluble in alcohol. Insoluble in acetone |
| Formula Weight | 115.03 |
| Grade | ACS |
| Odor | Odorless |
| Density | 1.803 g/cm3 |
| Chemical Name or Material | Ammonium dihydrogen phosphate |
Safety and Handling
| Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste. |
| EINECSNumber | 231-764-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only