missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

$306.99 - $306.99

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436791000
SDS View Documents
Thermo Scientific Chemicals
436791000
100 g Plastic bottle
Each for $306.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
phosphoric acid amine
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
CHEBI:62982
N.OP(O)(O)=O

Specifications

190.0°C
7722-76-1
100 g
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
LFVGISIMTYGQHF-UHFFFAOYSA-N
phosphoric acid amine
24402
115.03
Trace Metal Basis
Ammonium dihydrogen phosphate
White or Colorless
Crystals
99.999%
NH4H2PO4
15,541
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
N.OP(O)(O)=O
115.03
CHEBI:62982
99.999%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-764-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only