missing translation for 'onlineSavingsMsg'
Learn More

Ammonium copper(II) chloride dihydrate

CAS: 10060-13-6 | Cl4CuH12N2O2 | 277.454 g/mol

$167.34 - $672.58

Chemical Identifiers

CAS 10060-13-6
Molecular Formula Cl4CuH12N2O2
Molecular Weight (g/mol) 277.454
MDL Number MFCD00150384
InChI Key LLMDTWKBBWCWNN-UHFFFAOYSA-L
Synonym ammonium tetrachlorocuprate ii dihydrate, cupric ammonium chloride dihydrate, ammonium copper ii chloride dihydrate, diammonium tetrachlorocuprate ii dihydrate, ammonium chlorocuprate ii, copper ii ammonium chloride dihydrate, diammonium tetrachlorocuprate dihydrate, copper +2 diammonium chloride dihydrate, diammonium copper ii chloride dihydrate, ammonium tetrachlorocuprated ii dihydrate
PubChem CID 16210985
IUPAC Name diazanium;tetrachlorocopper(2-);dihydrate
SMILES [NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3912722
View Documents
Thermo Scientific Chemicals
03912722
100 g N/A
Only null left
 
AA3912736
View Documents
Thermo Scientific Chemicals
03912736
500 g N/A
Only null left
 
Description

Description

Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10060-13-6
277.454
LLMDTWKBBWCWNN-UHFFFAOYSA-L
16210985
[NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl
Cl4CuH12N2O2
MFCD00150384
ammonium tetrachlorocuprate ii dihydrate, cupric ammonium chloride dihydrate, ammonium copper ii chloride dihydrate, diammonium tetrachlorocuprate ii dihydrate, ammonium chlorocuprate ii, copper ii ammonium chloride dihydrate, diammonium tetrachlorocuprate dihydrate, copper +2 diammonium chloride dihydrate, diammonium copper ii chloride dihydrate, ammonium tetrachlorocuprated ii dihydrate
diazanium;tetrachlorocopper(2-);dihydrate

Specifications

110°C (decomposition)
10060-13-6
100 g
MFCD00150384
ammonium tetrachlorocuprate ii dihydrate, cupric ammonium chloride dihydrate, ammonium copper ii chloride dihydrate, diammonium tetrachlorocuprate ii dihydrate, ammonium chlorocuprate ii, copper ii ammonium chloride dihydrate, diammonium tetrachlorocuprate dihydrate, copper +2 diammonium chloride dihydrate, diammonium copper ii chloride dihydrate, ammonium tetrachlorocuprated ii dihydrate
LLMDTWKBBWCWNN-UHFFFAOYSA-L
diazanium;tetrachlorocopper(2-);dihydrate
16210985
1.993 g/mL
Blue
Crystals
Cl4CuH12N2O2
14,514
Soluble in water,acids and alcohol; Slightly soluble in ammonia.
[NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl
277.454
277.46 (241.43 Anhydrous)
Ammonium copper(II) chloride dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 239-690-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only