missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cobalt(II) phosphate monohydrate, 98%

CAS: 16827-96-6 | CoH6NO5P | 189.96 g/mol

Supplier:  Thermo Scientific Chemicals B2439318

Catalog No. AAB2439318

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium cobalt(II) phosphate monohydrate
50 g
NH4CoPO4·H2O
14,513
PNSIYUITZYKGRY-UHFFFAOYSA-L
azanium;cobalt(2+);phosphate;hydrate
25021442
98%
16827-96-6
CoH6NO5P
MFCD02683114
ammonium cobalt ii phosphate monohydrate, acmc-20alsz, ammonium cobalt phosphate hydrate, ammonium cobalt 2+ phosphate-water 1/1/1/1
[NH4+].O.[Co++].[O-]P([O-])([O-])=O
189.96
189.96 (171.94 Anhydrous)

Chemical Identifiers

16827-96-6
189.96
PNSIYUITZYKGRY-UHFFFAOYSA-L
25021442
[NH4+].O.[Co++].[O-]P([O-])([O-])=O
CoH6NO5P
MFCD02683114
ammonium cobalt ii phosphate monohydrate, acmc-20alsz, ammonium cobalt phosphate hydrate, ammonium cobalt 2+ phosphate-water 1/1/1/1
azanium;cobalt(2+);phosphate;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c

H302+H332-H317

EINECSNumber : 238-633-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only