missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cobalt(II) phosphate, anhydrous, 98%

Catalog No. p-7026744
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2293418 50 g
AAB2293430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2293418 Supplier Thermo Scientific Chemicals Supplier No. B2293418
Only null left

CAS: 14590-13-7 | CoH4NO4P | 171.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14590-13-7
Molecular Formula CoH4NO4P
Molecular Weight (g/mol) 171.94
MDL Number MFCD02683100
InChI Key FCXHHGTWMFGYPP-UHFFFAOYSA-L
Synonym ammonium cobaltous phosphate, ammonium cobalt ii phosphate, ammonium cobalt phosphate, kobaltammoniumphosphat, acmc-20alt0, ammonium cobalt orthophosphate, azanium cobalt 2+ phosphate, phosphoric acid, ammonium cobalt 2+ salt 1:1:1, ammonium cobalt ii phosphate, anhydrous, phosphoric acid,ammonium cobalt 2+ salt 1:1:1
PubChem CID 167120
IUPAC Name azanium;cobalt(2+);phosphate
SMILES [NH4+].[Co++].[O-]P([O-])([O-])=O

Specifications

Quantity 50 g
Linear Formula NH4CoPO4
Merck Index 14,513
Formula Weight 171.94
Percent Purity 98%
Chemical Name or Material Ammonium cobalt(II) phosphate, anhydrous
Hazard Category H302-H317-H350
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
EINECSNumber 238-633-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only