missing translation for 'onlineSavingsMsg'
Learn More

Ammonium citrate tribasic, 97+%

CAS: 3458-72-8 | C6H20N3O7+3 | 246.24 g/mol

$113.08 - $328.34

Chemical Identifiers

CAS 3458-72-8
Molecular Formula C6H20N3O7+3
Molecular Weight (g/mol) 246.24
MDL Number MFCD00036406
InChI Key YWYZEGXAUVWDED-UHFFFAOYSA-Q
Synonym ammonium citrate
PubChem CID 131675891
IUPAC Name azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium
SMILES C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1697322
View Documents
Thermo Scientific Chemicals
A1697322
100 g
Each for $113.08
Only null left
 
AAA1697336
View Documents
Thermo Scientific Chemicals
A1697336
500 g
Each for $328.34
Only null left
 
Description

Description

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

3458-72-8
246.24
YWYZEGXAUVWDED-UHFFFAOYSA-Q
131675891
C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N
C6H20N3O7+3
MFCD00036406
ammonium citrate
azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium

Specifications

3458-72-8
>97%
MFCD00036406
ammonium citrate
YWYZEGXAUVWDED-UHFFFAOYSA-Q
azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium
131675891
Ammonium citrate tribasic
∼180°C (decomposition)
C6H20N3O7+3
100 g
Soluble in water. Slightly soluble in alcohol.
C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N
246.24
243.22
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-394-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only