missing translation for 'onlineSavingsMsg'
Learn More

Ammonium citrate tribasic, 97+%

Catalog No. AAA1697322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1697322 100 g
AAA1697336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1697322 Supplier Thermo Scientific Chemicals Supplier No. A1697322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3458-72-8 | C6H20N3O7+3 | 246.24 g/mol

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3458-72-8
Molecular Formula C6H20N3O7+3
Molecular Weight (g/mol) 246.24
MDL Number MFCD00036406
InChI Key YWYZEGXAUVWDED-UHFFFAOYSA-Q
Synonym ammonium citrate
PubChem CID 131675891
IUPAC Name azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium
SMILES C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N

Specifications

Melting Point ∼180°C (decomposition)
Assay Percent Range >97%
Quantity 100 g
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 243.22
Chemical Name or Material Ammonium citrate tribasic
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-394-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only