missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Purified, Reagents

Catalog No. p-200021161
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337499 10 kg
01337498 3 kg
01337497 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337499 Supplier Labchem Inc Supplier No. C213200010KG
Only null left

Ammonium Chloride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym Ammonium Muriate
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Boiling Point 520°C
Melting Point 337.8°C
Color White
Physical Form Solid
Quantity 10 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.5274
Chemical Name or Material Ammonium Chloride, Purified