Learn More
Ammonium chloride, Puratronic™, 99.999% (metals basis)
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
$178.30 - $2670.90
Chemical Identifiers
| CAS | 12125-02-9 |
|---|---|
| Molecular Formula | ClH4N |
| Molecular Weight (g/mol) | 53.49 |
| MDL Number | MFCD00011420 |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
| PubChem CID | 25517 |
| ChEBI | CHEBI:31206 |
| SMILES | N.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1063218
|
Thermo Scientific Chemicals
01063218 |
50 g |
Each for $178.30
|
|
|||||
|
AA1063230
|
Thermo Scientific Chemicals
01063230 |
250 g |
Each for $744.96
|
|
|||||
|
AA10632A1
|
Thermo Scientific Chemicals
010632A1 |
1 kg |
Each for $2,670.90
|
|
|||||
Description
Ammonium chloride finds its use as a flux in preparing metals for galvanizing, soldering or tin coating. While mainly referred as a nitrogen source fertilizer, it is also used in cleaning tip of soldering iron, hair shampoo as a thickening agent, some batteries as electrolyte, tanning, textile printing, dyeing, and cleaning products. In laboratories it is used to produce cooling baths and for producing buffers. Along with certain metals, it is used for effecting certain reductions in organic synthesis. It can be used as a precursor to ammonium cyanate, which is employed in the Wöhler synthesis of urea. It facilitates several organic transformations in a mild and efficient manner - for example in Ulmann coupling, cross-aldol condensation, Claisen rearrangement, reduction, thia-Michael addition and synthesis of several monocyclic, fused or spiro bicyclic nitrogen heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| 25517 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| CHEBI:31206 |
Specifications
| 340° (sublimation) | |
| White | |
| 50 g | |
| ClH4N | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 25517 | |
| 53.49 | |
| 1.642 | |
| White powder | |
| Ammonium chloride |
| 12125-02-9 | |
| Powder | |
| 100.00% | |
| MFCD00011420 | |
| Highly soluble in water. Soluble in liquid ammonia,acetone,hydrazine,alcohol. Insoluble in diethyl ether and ethyl acetate. | |
| N.Cl | |
| 53.49 | |
| CHEBI:31206 | |
| Odorless | |
| Hygroscopic | |
| 1.53 g/cm3 |
Safety and Handling
P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
H302-H319-H335-H373
EINECSNumber : 235-186-4
RTECSNumber : BP4550000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only