missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Granular, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7157015
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600594 500 g Poly Bottle
18600595 2.5 kg Poly Bottle
18600596 12 kg Poly Pail
18600597 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600594 Supplier Spectrum Chemical Mfg Cor Supplier No. AM175500GM
Only null left

AM175, 12125-02-9, NH4Cl

Spectrum™ Chemical Ammonium Chloride, Granular, USP is used as an expectorant in cough medicine. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

pH 4.6-6.0
CAS Max % 100%
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade USP
Loss on Drying 0.5%
Packaging Poly Bottle