missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Granular, Reagent, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18600587
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600587 125 g Poly Bottle
18600588 500 g Poly Bottle
18600590 2.5 kg Poly Bottle
18600589 6 x 500 g Poly Bottle
18600591 4 x 2.5 kg Poly Bottle
18600592 12 kg Poly Pail
18600593 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18600587 Supplier Spectrum Chemical Mfg Cor Supplier No. A1165125GM
Only null left

A1165, 12125-02-9, NH4Cl

Spectrum™ Chemical Ammonium Chloride, Granular, Reagent, ACS is an inorganic compound, that presents itself as a white crystalline salt that is highly soluble in water. It occurs from the reaction of hydrochloric acid and ammonia. It has uses in medicine as an expectorant in cough medicine, in metalwork as a flux in preparing metals to be tin coated as well as in laboratories to produce cooling baths at low temperatures. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Chloride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

pH 4.5-5.5
CAS Max % 100%
Quantity 125 g
Percent Purity 99.5%
Grade ACS
Packaging Poly Bottle
Residue after Ignition 0.01%