missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, ACS, 99.5% min

Catalog No. AA40193A7
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40193A7 5 kg
AA40193A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40193A7 Supplier Thermo Scientific Chemicals Supplier No. 040193A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacturing of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340° (sublimation)
Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99.5% min
Solubility Information Soluble in liquid ammonia,acetone,hydrazine and alcohol. Insoluble in diethyl ether and ethyl acetate.
IUPAC Name amine hydrochloride
Formula Weight 53.49
Grade ACS Reagent
Refractive Index 1.642
Sensitivity Hygroscopic
Density 1.53
Chemical Name or Material Ammonium chloride
Show More Show Less
Hazard Category H302-H319-H335-H373
Precautionary Statement P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
EINECSNumber 235-186-4
RTECSNumber BP4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only