missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, ≥99.5% (ACS Guaranteed Reagent), MilliporeSigma™
Specifications
Specifications
| Boiling Point | 520°C |
| Melting Point | 335°C |
| Physical Form | Crystals |
| Quantity | 500 g |
| Assay Percent Range | ≥99.5% (silver nitrate titration) |
| Linear Formula | NH4Cl |
| Formula Weight | 53.49 |
| Percent Purity | ≥99.5% |
| Grade | ACS |
| Ignition Point | >400°C |
| Show More |
Safety and Handling
| Recommended Storage | Store without restrictions |