missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 99.7%, MP Biomedicals™

Numéro de catalogue. ICN15010991
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ICN15010991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. ICN15010991 Fournisseur MP Biomedicals Inc Code fournisseur 0215010991
Il en reste null

Identifiants chimiques

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Spécifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 0.997
Formula Weight 53.5
Chemical Name or Material Ammonium chloride