Learn More
Ammonium chloride, 99.6%, ACS reagent
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Supplier: Thermo Scientific Chemicals 423280010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
| Ammonium chloride | |
| 12125-02-9 | |
| 4.5 to 5.5 (5% soln. at 25°C) | |
| 1 kg | |
| ClH4N | |
| MFCD00011420 | |
| 15, 506 | |
| Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate | |
| N.Cl | |
| 53.49 | |
| CHEBI:31206 | |
| 99.6% | |
| Plastic bottle | |
| 1.5200g/mL |
| 340.0°C | |
| White | |
| Crystalline Powder | |
| 99.6% | |
| NH3·HCl | |
| 15,13 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 25517 | |
| 53.49 | |
| ACS Reagent | |
| 0.01% max |
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| 25517 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| CHEBI:31206 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d
GHS Signal Word: Warning
EINECSNumber : 235-186-4
RUO – Research Use Only