Learn More
Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
$127.86 - $127.86
Chemical Identifiers
| CAS | 12125-02-9 |
|---|---|
| Molecular Formula | ClH4N |
| Molecular Weight (g/mol) | 53.49 |
| MDL Number | MFCD00011420 |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
| PubChem CID | 25517 |
| ChEBI | CHEBI:31206 |
| SMILES | N.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC396400010
|
Thermo Scientific Chemicals
396400010 |
1 kg | Plastic Bottle |
Each for $127.86
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| 25517 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| CHEBI:31206 |
Specifications
| 340.0°C | |
| White | |
| 1 kg | |
| ClH4N | |
| MFCD00011420 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| azanium;chloride | |
| 25517 | |
| 53.49 | |
| Extra Pure | |
| 1.52 | |
| Ammonium chloride |
| 12125-02-9,7732-18-5 | |
| Crystalline Powder | |
| 99.0 to 101% (Argentometry) | |
| NH4Cl | |
| 15, 506 | |
| Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and ethyl,acetate | |
| N.Cl | |
| 53.49 | |
| CHEBI:31206 | |
| ≥99% | |
| Plastic Bottle | |
| 1.5200g/mL |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha
GHS Signal Word: Warning
EINECSNumber : 235-186-4
RUO – Research Use Only