missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 98+%

Catalog No. p-7022293
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500030 250 g
AAA150000B 1000 g
AAA150000I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500030 Supplier Thermo Scientific Chemicals Supplier No. A1500030

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.

Solubility
Soluble in water, methanol, liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.

Notes
Hygroscopic. Incompatible with silver nitrate and strong oxidizers.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340°C (sublimation)
Quantity 250 g
Linear Formula NH4Cl
Beilstein 4371014
Merck Index 14,509
Solubility Information Soluble in water,methanol,liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.
IUPAC Name azanium;chloride
Formula Weight 53.49
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.642
Sensitivity Hygroscopic
Density 1.53
Chemical Name or Material Ammonium chloride
Show More Show Less
Hazard Category H302-H319-H335-H373
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
EINECSNumber 235-186-4
RTECSNumber BP4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only