missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, 1.15 g/L, pH 7.20, for BOD, Ricca Chemical
For BOD Testing
Supplier: Ricca Chemical Company 62316
Specifications
| Clear colorless | |
| 7.2 | |
| Liquid | |
| MFCD00011420 | |
| Natural Poly Bottle | |
| amine hydrochloride | |
| 500 mL |
| 1.15 g/L | |
| 500 mL | |
| ClH4N | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| N.Cl | |
| 53.49 | |
| Ammonium Chloride |