missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure

Catalog No. AC206310050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206310050 5 g Glass bottle
AC206311000 100 g Glass bottle
AC206315000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC206310050 Supplier Thermo Scientific Chemicals Supplier No. 206310050

CAS: 10378-47-9 | H16CeN4O16S4·2H2O | 632.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10378-47-9
Molecular Formula H16CeN4O16S4·2H2O
Molecular Weight (g/mol) 632.54
MDL Number MFCD00149417
Synonym Ceric ammonium sulfate

Specifications

Melting Point 130.0°C
Color Orange
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99+%
Linear Formula (NH4)4Ce(SO4)4·2H2O
Fieser 08,80
Solubility Information Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol
Formula Weight 632.54
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Ammonium cerium(IV) sulfate dihydrate, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only