missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(Iv) Sulfate Dihydrate, ≥98.0% (RT), puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7120170
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045345 100 g Poly Bottle
60045346 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045345 Supplier Honeywell Chemicals Supplier No. 22269100G
Only null left

Puriss. p.a., ACS Reagent,≥98.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10378-47-9
MDL Number MFCD00149417

Specifications

Quantity 100 g
Linear Formula Ce(NH4)4(SO4)4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 632.55g/mol
Percent Purity ≥98.0% (RT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Chemical Name or Material Ammonium Cerium(Iv) Sulfate Dihydrate