missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(IV) Nitrate 98.0+%, TCI America™

Catalog No. p-7125182
Change view
Click to view available options
Quantity:
50 g
500 g
Catalog No. Quantity
C180650G 50 g
C1806500G 500 g
2 options

Catalog No. C180650G

Supplier: TCI America C180650G

Only null left

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
Synonym Ceric Ammonium Nitrate, CAN, Cerium(IV) Diammonium Nitrate, Diammonium Cerium(IV) Nitrate

Specifications

Color Red-Yellow
Physical Form Crystalline Powder
Quantity 50 g
UN Number 1477
Formula Weight 548.22
Percent Purity ≥98.0% (T)
Chemical Name or Material Ammonium Cerium(IV) Nitrate