Learn More
Ammonium cerium(IV) nitrate, pure, Thermo Scientific™
Oxidation reagent
Chemical Identifiers
| CAS | 16774-21-3 |
|---|---|
| Molecular Formula | H8CeN8O18 |
| Molecular Weight (g/mol) | 548.22 |
| Synonym | Ceric ammonium nitrate, CAN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC212360010
|
Thermo Scientific Chemicals
212360010 |
1kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 16774-21-3 | |
| H8CeN8O18 | |
| 548.22 | |
| Ceric ammonium nitrate, CAN |
Specifications
| 16774-21-3 | |
| H8CeN8O18 | |
| 01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68 | |
| Ceric ammonium nitrate, CAN | |
| 548.22 | |
| Pure | |
| Ammonium cerium(IV) nitrate |
| 1kg | |
| (NH4)2Ce(NO3)6 | |
| 15,1991 | |
| Solubility in water: 1410g/L (20°C). Other solubilities: insoluble in fat, soluble in alcohol | |
| 548.22 | |
| 2% max. |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 240-827-6
RUO â Research Use Only