missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18600573
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600573 500 mL Amber Glass Bottle
18600572 100 mL Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600573 Supplier Spectrum Chemical Mfg Cor Supplier No. A302500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A-302, 506-87-6

Spectrum™ Chemical Ammonium Carbonate TS, (U.S.P. Test Solution) is a solution, sometimes designated with TS or Test Solution, that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 506-87-6 , 1336-21-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

CAS Max % 0.1%, 99.9%
Physical Form Liquid
Quantity 500 mL
Grade USP
Packaging Amber Glass Bottle
Chemical Name or Material Ammonium Carbonate TS