missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, puriss., Solstice

Catalog No. 6001897
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001897 1 kg
6001898 2.5 kg
60044688 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001897 Supplier Solstice Supplier No. 112041KG

Puriss., meets analytical specification of NF, Ph. Franc., FCC

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Quantity 1 kg
Linear Formula (NH4)2CO3
Beilstein 4253487
Formula Weight 96.09g/mol
Grade puriss.
Residue after Ignition ≤0.05% (as SO4)
Density Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium Carbonate, Meets analytical specification of NF, Ph.Franc.,FCC
Hazard Statement P301 + P312 + P330
Precautionary Statement H302
EINECSNumber 208-058-0
RTECSNumber BP1925000