missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Purified, Reagents

Catalog No. p-200020909
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337501 2.5 kg
01337500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337501 Supplier Labchem Inc Supplier No. C21315002.5KG
Only null left

Ammonium Carbonate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym Smelling salts
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

Boiling Point 60°C
Melting Point 58°C
Color White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.5
Chemical Name or Material Ammonium Carbonate, Purified