missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Purified, Reagents

Catalog No. 01337501
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337501 2.5 kg
01337500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337501 Supplier Labchem Inc Supplier No. C21315002.5KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Carbonate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym Smelling salts
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

Boiling Point 60°C
Melting Point 58°C
Color White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.5
Chemical Name or Material Ammonium Carbonate, Purified