missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Powder, Technical, Spectrum™ Chemical

Catalog No. 18608604
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608604 50 kg Poly Pail
18608603 500 g Poly Pail
18608602 2.5 kg Poly Pail
18608601 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608604 Supplier Spectrum Chemical Mfg Cor Supplier No. A115450KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-87-6

Specifications

CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 50 kg
Grade Technical
Packaging Poly Pail
Chemical Name or Material Ammonium Carbonate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.