missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, NH3 ca 30%

Catalog No. AAA139550I
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA139550I 5000 g
AAA139550B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA139550I Supplier Thermo Scientific Chemicals Supplier No. A139550I

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium carbonate is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It may be used as a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements and in the synthesis of ZnO nanoparticles. It may also be used as an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass . It was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Melting Point 60°C (sublimation)
Quantity 5000 g
Linear Formula (NH4)2CO3
Beilstein 3627235
Merck Index 14,508
Solubility Information Soluble in water.
Formula Weight 96.09
Odor Ammonia-like
Assay NH3 ≈30%
Chemical Name or Material Ammonium carbonate
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-058-0
RTECSNumber BP1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only