missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, MP Biomedicals™

Catalog No. ICN15010880
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15010880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15010880 Supplier MP Biomedicals Inc Supplier No. 0215010880

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10361-29-2
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym hartshorn salt, bis soda triamine, ammonium carbonate, acs, bis carbonic acid triamine
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 96.09
Chemical Name or Material Ammonium carbonate