missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Chips, Purified, Spectrum™ Chemical

Catalog No. 18608594
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608594 500 g Poly Pail
18608592 2.5 kg Poly Pail
18608591 12 kg Poly Pail
18608593 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608594 Supplier Spectrum Chemical Mfg Cor Supplier No. A1152500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-87-6

Specifications

CAS Max % 1
Physical Form Chips
Quantity 500 g
Grade Purified
Packaging Poly Pail
Chemical Name or Material Ammonium Carbonate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.