missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071850
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002056 100 g
6002060 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002056 Supplier Honeywell Chemicals Supplier No. 207861100G
Only null left

ACS Reagent,≥30.0% NH3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Quantity 100 g
Linear Formula (NH4)2CO3
Beilstein 4253487
Formula Weight 96.09g/mol
Percent Purity ≥30% (NH3 basis)
Grade ACS Reagent
Density Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium Carbonate
Hazard Statement P301 + P312 + P330
Precautionary Statement H302
EINECSNumber 208-058-0
RTECSNumber BP1925000