missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, ACS, NH{3} 30.0+%

Catalog No. AA36229A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36229A3 2 kg
AA3622936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36229A3 Supplier Thermo Scientific Chemicals Supplier No. 036229A3

In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue | CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds

Chemical Identifiers

CAS 506-87-6
MDL Number MFCD00010890

Specifications

Melting Point 60°C (sublimation)
Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range NH3 30.0+%
Molecular Formula CH8N2O3
Beilstein 3627235
Merck Index 14,508
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
Solubility Information Slowly soluble in water (1:4 portion); Decomposes in hot water.
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
IUPAC Name diazanium;carbonate
Molecular Weight (g/mol) 96.086
PubChem CID 517111
Formula Weight 96.09
Grade ACS
Concentration or Composition (by Analyte or Components) NH3 ≥30.0%
Odor Ammonia-like
Sensitivity Hygroscopic
Chemical Name or Material Ammonium carbonate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-058-0
RTECSNumber BP1925000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only