missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Carbonate ACS AR, Macron Fine Chemicals™
Specifications
Specifications
| CAS | 10361-29-2 |
| Quantity | 500g |
| Molecular Formula | CH8N2O3 |
| Synonym | hartshorn salt, bis soda triamine, ammonium carbonate, acs, bis carbonic acid triamine |
| InChI Key | PRKQVKDSMLBJBJ-UHFFFAOYSA-N |
| SMILES | C(=O)([O-])[O-].[NH4+].[NH4+] |
| IUPAC Name | diazanium;carbonate |
| Molecular Weight (g/mol) | 96.086 |
| PubChem CID | 517111 |
| Grade | ACS |
| Show More |