missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbamate 97.0+%, TCI America™

Catalog No. p-7124184
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C002725G 25 g
C0027500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C002725G Supplier TCI America Supplier No. C002725G
Only null left

Chemical Identifiers

CAS 1111-78-0
Molecular Formula CH6N2O2
Molecular Weight (g/mol) 78.071
MDL Number MFCD00013010
InChI Key BVCZEBOGSOYJJT-UHFFFAOYSA-N
Synonym ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate
PubChem CID 517232
IUPAC Name azanium;carbamate
SMILES C(=O)(N)[O-].[NH4+]

Specifications

Melting Point 59°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 78.07
Percent Purity ≥97.0% (T)
Chemical Name or Material Ammonium Carbamate
EINECSNumber (2)-3778
RTECSNumber EY8575000
TSCA Yes
Recommended Storage Refrigerator