missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bromide, Granular, Spectrum™ Chemical

Catalog No. 18608589
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608589 2.5 kg Poly Pail
18608585 25 g Poly Bottle
18608587 500 g Poly Bottle
18608586 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608589 Supplier Spectrum Chemical Mfg Cor Supplier No. A11512.5KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12124-97-9

Specifications

pH 4.5 to 6.0
CAS Max % 1
Quantity 2.5 kg
Grade Reagent
Packaging Poly Pail
Residue after Ignition 0.0001

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.