missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bromide, Granular, Spectrum™ Chemical
Specifications
Specifications
| pH | 4.5 to 6.0 |
| CAS Max % | 1 |
| Quantity | 25 kg |
| Grade | Reagent |
| Packaging | Poly Pail |
| Residue after Ignition | 0.0001 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.