missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, ACS, 99.0% min

Catalog No. AA14257A3
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14257A3 2 kg
AA1425714 25 g
AA1425722 100 g
AA1425736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA14257A3 Supplier Thermo Scientific Chemicals Supplier No. 014257A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woodsAmmonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452° (sublimation)
Color Colorless
Physical Form Solid
Quantity 2 kg
Assay Percent Range 99% min.
Solubility Information Freely soluble in water,methanol,ethanol,and acetone. Slightly soluble in ether. Practically insoluble in ethyl acetateSoluble in water,methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 97.97
Grade ACS
Sensitivity Hygroscopic
Density 2.429
Chemical Name or Material Ammonium bromide
Show More Show Less
Hazard Category H319
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 235-183-8
RTECSNumber BO9155000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only