missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, 99%

Catalog No. aaa162580c
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10,000 g
2500 g
500 g
Catalog No. Quantity
AAA162580C 10,000 g
AAA1625836 500 g
AAA162580E 2500 g
3 options

Catalog No. AAA162580C

Supplier: Thermo Scientific Chemicals A162580C

Only null left

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Ammonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

Solubility
Soluble in water, methanol, ethanol and acetone. Slightly soluble in ether.

Notes
Hygroscopic and air sensitive. Incompatible with strong oxidizing agents, acids, bases, heavy metals, silver salts and potassium.

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452°C (sublimation)
Quantity 10,000 g
Merck Index 14,505
Solubility Information Soluble in water,methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 97.97
Percent Purity 99%
Sensitivity Hygroscopic
Density 2.429
Chemical Name or Material Ammonium bromide
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 235-183-8
RTECSNumber BO9155000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only