missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, 98%, extra pure

Catalog No. AC450650100
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450650100 10 kg Plastic drum
AC450650010 1 kg Plastic bottle
AC450650025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450650100 Supplier Thermo Scientific Chemicals Supplier No. 450650100

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Boiling Point 235.0°C
Melting Point 452.0°C
Quantity 10 kg
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula NH4Br
Merck Index 15,502
Solubility Information Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate
Formula Weight 97.94
Percent Purity 98%
Grade Extra Pure
Packaging Plastic drum
Chemical Name or Material Ammonium bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 235-183-8

RUO – Research Use Only